Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191838.1 | NP_001178767.1 | 4 - 5 | 495 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |