Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014022.1 | NP_001014044.1 | 1 - 2 | 94 | 76 | |
XM_006230034.2 | XP_006230096.1 | 1 - 2 | 351 | 76 | ||
XM_006230036.2 | XP_006230098.1 | 1 - 2 | 351 | 76 | ||
XM_006230041.2 | XP_006230103.1 | 1 - 2 | 351 | 76 | ||
XM_008759725.1 | XP_008757947.1 | 1 - 2 | 351 | 76 | ||
GenBank mRNA | BC090330.1 | - | 1 - 2 | 94 | 7300759 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |