Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 5 - 6 | 647 | 73 | |
GenBank mRNA | AF000578.2 | - | 5 - 6 | 647 | 73 | |
BC087007.1 | - | 5 - 6 | 584 | 73 | ||
BC161839.1 | - | 5 - 6 | 611 | 9101355 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |