Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244572.2 | XP_006244634.1 | 1 - 2 | 281 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |