Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729998.4 | XP_002730044.2 | 3 - 4 | 496 | 65 | |
XM_006244572.2 | XP_006244634.1 | 3 - 4 | 499 | 65 | ||
XM_006244573.2 | XP_006244635.1 | 4 - 5 | 361 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |