Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053537.2 | NP_445989.2 | 8 - 9 | 1393 | 121 | |
GenBank mRNA | AY816233.1 | - | 8 - 9 | 1393 | 121 | |
L27651.1 | - | 8 - 9 | 1453 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |