Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025707.1 | NP_001020878.1 | 2 - 3 | 713 | 67 | |
XM_006244440.2 | XP_006244502.1 | 3 - 4 | 1642 | 67 | ||
XM_006244442.1 | XP_006244504.1 | 3 - 4 | 632 | 67 | ||
XM_006244446.2 | XP_006244508.1 | 3 - 4 | 1903 | 67 | ||
GenBank mRNA | BC099102.1 | - | 2 - 3 | 713 | 7460021 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |