Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013178.2 | NP_001013196.1 | 1 - 2 | 121 | 108 | |
NM_001277367.1 | NP_001264296.1 | 1 - 2 | 121 | 108 | ||
GenBank mRNA | BC083734.1 | - | 1 - 2 | 103 | 7191561 | 108 |
FN804985.1 | - | 1 - 2 | 83 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |