Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106898.1 | NP_001100368.1 | 24 - 25 | 3498 | 59 | |
XM_008766928.1 | XP_008765150.1 | 8 - 9 | 1189 | 59 | ||
XM_008766929.1 | XP_008765151.1 | 9 - 10 | 1082 | 59 | ||
XM_008766930.1 | XP_008765152.1 | 9 - 10 | 2503 | 59 | ||
XM_008766931.1 | XP_008765153.1 | 9 - 10 | 1319 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |