Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133284.2 | NP_579818.1 | 4 - 5 | 522 | 88 | |
GenBank mRNA | X04644.1 | - | 4 - 5 | 522 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |