Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108205.1 | NP_001101675.1 | 1 - 2 | 283 | 85 | |
XM_006244538.2 | XP_006244600.1 | 1 - 2 | 805 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |