Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750667.3 | XP_003750715.3 | 8 - 9 | 1101 | 109 | |
XM_003750668.3 | XP_003750716.3 | 9 - 10 | 1183 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |