Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110138.1 | NP_001103608.1 | 3 - 4 | 1035 | 89 | |
GenBank mRNA | BC161933.1 | - | 3 - 4 | 1054 | 9022122 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |