Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025693.1 | NP_001020864.1 | 3 - 4 | 481 | 67 | |
GenBank mRNA | BC098690.1 | - | 3 - 4 | 481 | 7386103 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |