Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107760.1 | NP_001101230.1 | 8 - 9 | 1559 | 68 | |
XM_006234765.2 | XP_006234827.1 | 8 - 9 | 1570 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |