Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004265.2 | NP_001004265.2 | 5 - 6 | 1149 | 79 | |
XM_006234763.1 | XP_006234825.1 | 6 - 7 | 1161 | 79 | ||
XM_006234764.2 | XP_006234826.1 | 6 - 7 | 1087 | 79 | ||
XM_008762100.1 | XP_008760322.1 | 6 - 7 | 1161 | 79 | ||
GenBank mRNA | BC097253.1 | - | 5 - 6 | 1149 | 7459422 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |