Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107812.2 | NP_001101282.2 | 7 - 8 | 861 | 77 | |
XM_008761908.1 | XP_008760130.1 | 6 - 7 | 706 | 77 | ||
GenBank mRNA | BC167092.1 | - | 7 - 8 | 788 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |