Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Api5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127379.1 | NP_001120851.1 | 6 - 7 | 814 | 124 | |
XM_006234586.1 | XP_006234648.1 | 6 - 7 | 907 | 124 | ||
GenBank mRNA | BC162003.1 | - | 6 - 7 | 814 | 9089120 | 124 |
DQ480749.1 | - | 6 - 7 | 891 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |