Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106504.1 | NP_001099974.1 | 4 - 5 | 388 | 76 | |
XM_008762157.1 | XP_008760379.1 | 4 - 5 | 331 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |