Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyltl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199107.1 | NP_954538.1 | 4 - 5 | 365 | 86 | |
XM_006234562.2 | XP_006234624.1 | 3 - 4 | 1686 | 86 | ||
XM_006234564.2 | XP_006234626.1 | 4 - 5 | 425 | 86 | ||
XM_006234566.2 | XP_006234628.1 | 4 - 5 | 425 | 86 | ||
XM_008762024.1 | XP_008760246.1 | 3 - 4 | 498 | 86 | ||
XM_008762025.1 | XP_008760247.1 | 3 - 4 | 613 | 86 | ||
GenBank mRNA | BC061762.1 | - | 4 - 5 | 365 | 5621056 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |