Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107778.1 | NP_001101248.1 | 8 - 9 | 998 | 82 | |
XM_006235027.2 | XP_006235089.1 | 9 - 10 | 1153 | 82 | ||
XM_006235028.2 | XP_006235090.1 | 8 - 9 | 1033 | 82 | ||
XM_008762178.1 | XP_008760400.1 | 8 - 9 | 2074 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |