Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033707.1 | NP_001028879.1 | 6 - 7 | 605 | 75 | |
XM_006234529.2 | XP_006234591.1 | 6 - 7 | 710 | 75 | ||
XM_006234530.2 | XP_006234592.1 | 6 - 7 | 709 | 75 | ||
XM_006234531.2 | XP_006234593.1 | 6 - 7 | 709 | 75 | ||
GenBank mRNA | BC101917.1 | - | 6 - 7 | 605 | 7935829 | 75 |
FQ218185.1 | - | 6 - 7 | 677 | 75 | ||
FQ228393.1 | - | 6 - 7 | 672 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |