Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017092.1 | NP_058788.1 | 12 - 13 | 1571 | 70 | |
GenBank mRNA | D37880.1 | - | 12 - 13 | 1571 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |