Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170594.1 | NP_001164065.1 | 14 - 15 | 5499 | 130 | |
XM_008762095.1 | XP_008760317.1 | 17 - 18 | 5685 | 130 | ||
XM_008762096.1 | XP_008760318.1 | 17 - 18 | 5915 | 130 | ||
XM_008762097.1 | XP_008760319.1 | 17 - 18 | 5728 | 130 | ||
XM_008762098.1 | XP_008760320.1 | 15 - 16 | 5598 | 130 | ||
XM_008762099.1 | XP_008760321.1 | 17 - 18 | 5672 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |