Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077429.1 | NP_001070897.1 | 8 - 9 | 609 | 102 | |
XM_006234377.2 | XP_006234439.1 | 8 - 9 | 691 | 102 | ||
XM_006234378.2 | XP_006234440.1 | 9 - 10 | 712 | 102 | ||
XM_006234379.2 | XP_006234441.1 | 8 - 9 | 739 | 102 | ||
XM_006234380.2 | XP_006234442.1 | 8 - 9 | 811 | 102 | ||
XM_006234381.2 | XP_006234443.1 | 8 - 9 | 904 | 102 | ||
XM_006234382.2 | XP_006234444.1 | 9 - 10 | 832 | 102 | ||
XM_006234385.2 | XP_006234447.1 | 8 - 9 | 693 | 102 | ||
XM_008761909.1 | XP_008760131.1 | 8 - 9 | 1788 | 102 | ||
XM_008761910.1 | XP_008760132.1 | 8 - 9 | 693 | 102 | ||
GenBank mRNA | BC126100.1 | - | 8 - 9 | 609 | 7132654 | 102 |
FQ218047.1 | - | 7 - 8 | 488 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |