Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012088.1 | NP_001012088.1 | 8 - 9 | 834 | 111 | |
XM_006234567.2 | XP_006234629.1 | 8 - 9 | 940 | 111 | ||
GenBank mRNA | BC089938.1 | - | 8 - 9 | 834 | 7310024 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |