Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013078.1 | NP_001013096.1 | 7 - 8 | 993 | 99 | |
XM_006234734.2 | XP_006234796.1 | 7 - 8 | 1031 | 99 | ||
XM_006234735.2 | XP_006234797.1 | 7 - 8 | 1028 | 99 | ||
XM_008762090.1 | XP_008760312.1 | 8 - 9 | 974 | 99 | ||
GenBank mRNA | BC082080.1 | - | 7 - 8 | 993 | 7133293 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |