Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012090.1 | NP_001012090.1 | 2 - 3 | 256 | 105 | |
GenBank mRNA | BC079122.1 | - | 2 - 3 | 256 | 7126707 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |