Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016988.2 | NP_058684.2 | 1 - 2 | 149 | 69 | |
XM_006234500.1 | XP_006234562.1 | 2 - 3 | 1071 | 69 | ||
GenBank mRNA | BC081823.1 | - | 1 - 2 | 120 | 7107570 | 69 |
CB720349.1 | - | 1 - 2 | 149 | 69 | ||
M27893.1 | - | 1 - 2 | 118 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |