Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133289.1 | NP_579823.1 | 17 - 18 | 3471 | 66 | |
GenBank mRNA | AF000368.1 | - | 17 - 18 | 3471 | 66 | |
U79568.1 | - | 17 - 18 | 3471 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |