Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107762.1 | NP_001101232.1 | 3 - 4 | 579 | 101 | |
XM_006234768.2 | XP_006234830.1 | 3 - 4 | 410 | 101 | ||
XM_006234769.2 | XP_006234831.1 | 3 - 4 | 410 | 101 | ||
XM_008762101.1 | XP_008760323.1 | 3 - 4 | 410 | 101 | ||
XM_008762102.1 | XP_008760324.1 | 3 - 4 | 410 | 101 | ||
GenBank mRNA | BC161947.1 | - | 3 - 4 | 355 | 9025380 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |