Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 4 - 5 | 629 | 71 | |
GenBank mRNA | BC062019.1 | - | 4 - 5 | 539 | 5620971 | 71 |
D83522.1 | - | 4 - 5 | 503 | 71 | ||
FQ225567.1 | - | 3 - 4 | 288 | 71 | ||
FQ225947.1 | - | 3 - 4 | 288 | 71 | ||
FQ226273.1 | - | 3 - 4 | 288 | 71 | ||
FQ232987.1 | - | 3 - 4 | 288 | 71 | ||
FQ233836.1 | - | 3 - 4 | 288 | 71 | ||
U77918.1 | - | 4 - 5 | 629 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |