Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 2 - 3 | 805 | 96 | |
XM_006234290.2 | XP_006234352.1 | 3 - 4 | 1040 | 96 | ||
GenBank mRNA | AB040674.1 | - | 2 - 3 | 694 | 96 | |
BC101886.1 | - | 2 - 3 | 808 | 7929522 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |