Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271266.1 | NP_001258195.1 | 4 - 5 | 1477 | 83 | |
GenBank mRNA | AF127390.1 | - | 4 - 5 | 1477 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |