Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022502.2 | NP_071947.1 | 3 - 4 | 365 | 83 | |
GenBank mRNA | BC126089.1 | - | 3 - 4 | 365 | 8361877 | 83 |
FQ213474.1 | - | 3 - 4 | 377 | 83 | ||
FQ213911.1 | - | 377 | 83 | |||
L34262.1 | - | 363 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |