Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005903.1 | NP_001005903.1 | 6 - 7 | 671 | 79 | |
XM_006239171.2 | XP_006239233.1 | 7 - 8 | 1723 | 79 | ||
XM_006239172.2 | XP_006239234.1 | 7 - 8 | 1720 | 79 | ||
XM_006239174.2 | XP_006239236.1 | 6 - 7 | 1650 | 79 | ||
XM_006239175.2 | XP_006239237.1 | 6 - 7 | 1646 | 79 | ||
XM_006239177.2 | XP_006239239.1 | 6 - 7 | 700 | 79 | ||
GenBank mRNA | BC083861.1 | - | 6 - 7 | 671 | 7126848 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |