Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108691.1 | NP_001102161.1 | 4 - 5 | 796 | 77 | |
XM_006239253.2 | XP_006239315.1 | 4 - 5 | 796 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |