Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048184.1 | NP_001041649.1 | 2 - 3 | 477 | 102 | |
GenBank mRNA | BC105622.1 | - | 2 - 3 | 477 | 7454011 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |