Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080783.2 | NP_542961.2 | 6 - 7 | 670 | 86 | |
XM_006239134.2 | XP_006239196.1 | 6 - 7 | 1635 | 86 | ||
XM_008764197.1 | XP_008762419.1 | 6 - 7 | 901 | 86 | ||
GenBank mRNA | BC097293.1 | - | 6 - 7 | 670 | 7443707 | 86 |
X53949.1 | - | 5 - 6 | 648 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |