Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113784.1 | NP_001107256.1 | 2 - 3 | 211 | 62 | |
XM_006238886.2 | XP_006238948.1 | 2 - 3 | 471 | 62 | ||
XM_006238887.2 | XP_006238949.1 | 2 - 3 | 471 | 62 | ||
GenBank mRNA | BC158755.1 | - | 2 - 3 | 211 | 9103162 | 62 |
FQ212480.1 | - | 2 - 3 | 223 | 62 | ||
FQ215854.1 | - | 2 - 3 | 229 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |