Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024386.1 | NP_077362.1 | 3 - 4 | 268 | 62 | |
GenBank mRNA | BC061797.1 | - | 3 - 4 | 270 | 5598033 | 62 |
FQ209392.1 | - | 3 - 4 | 266 | 62 | ||
FQ209616.1 | - | 3 - 4 | 272 | 62 | ||
FQ234382.1 | - | 3 - 4 | 266 | 62 | ||
Y10054.1 | - | 3 - 4 | 268 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |