Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053305.1 | NP_445757.1 | 1 - 2 | 184 | 60 | |
GenBank mRNA | AF127389.1 | - | 1 - 2 | 184 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel Rattus norvegicus Probe spans exons |