Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100709.1 | NP_001094179.1 | 10 - 11 | 1209 | 75 | |
XM_006238932.2 | XP_006238994.1 | 11 - 12 | 1376 | 75 | ||
XM_006238933.2 | XP_006238995.1 | 11 - 12 | 1338 | 75 | ||
XM_008764140.1 | XP_008762362.1 | 11 - 12 | 1394 | 75 | ||
GenBank mRNA | BC099218.1 | - | 6 - 7 | 813 | 75 | |
BC160881.1 | - | 10 - 11 | 1209 | 7381544 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |