Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025624.2 | NP_001020795.1 | 4 - 5 | 302 | 78 | |
GenBank mRNA | BC099178.1 | - | 4 - 5 | 268 | 7383003 | 78 |
BC099815.1 | - | 4 - 5 | 277 | 7929654 | 78 | |
CB614313.1 | - | 4 - 5 | 311 | 78 | ||
FQ213300.1 | - | 4 - 5 | 270 | 78 | ||
FQ213727.1 | - | 4 - 5 | 261 | 78 | ||
FQ214234.1 | - | 2 - 3 | 97 | 78 | ||
FQ214394.1 | - | 4 - 5 | 261 | 78 | ||
FQ214422.1 | - | 4 - 5 | 272 | 78 | ||
FQ215102.1 | - | 4 - 5 | 278 | 78 | ||
FQ216264.1 | - | 4 - 5 | 296 | 78 | ||
FQ216489.1 | - | 4 - 5 | 233 | 78 | ||
FQ217912.1 | - | 4 - 5 | 272 | 78 | ||
FQ219745.1 | - | 4 - 5 | 337 | 78 | ||
FQ219836.1 | - | 4 - 5 | 299 | 78 | ||
FQ220601.1 | - | 4 - 5 | 313 | 78 | ||
FQ221300.1 | - | 4 - 5 | 243 | 78 | ||
FQ229179.1 | - | 4 - 5 | 261 | 78 | ||
FQ229184.1 | - | 4 - 5 | 235 | 78 |
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 268 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |