Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025736.1 | NP_001020907.1 | 2 - 3 | 209 | 91 | |
GenBank mRNA | BC099173.1 | - | 2 - 3 | 209 | 7382617 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |