Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029923.1 | NP_001025094.1 | 3 - 4 | 490 | 82 | |
GenBank mRNA | AB299031.1 | - | 3 - 4 | 393 | 82 | |
BC098048.1 | - | 3 - 4 | 490 | 7457181 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |