Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029923.1 | NP_001025094.1 | 5 - 6 | 664 | 92 | |
GenBank mRNA | AB299031.1 | - | 5 - 6 | 567 | 92 | |
BC098048.1 | - | 5 - 6 | 664 | 7457181 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |