Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF000942.1 | - | 1 | 400 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |