Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008524.1 | NP_001008524.1 | 2 - 3 | 205 | 94 | |
XM_006239225.1 | XP_006239287.1 | 2 - 3 | 485 | 94 | ||
GenBank mRNA | BC086409.1 | - | 2 - 3 | 205 | 7306579 | 94 |
FQ218827.1 | - | 2 - 3 | 229 | 94 | ||
FQ219610.1 | - | 2 - 3 | 257 | 94 | ||
FQ220269.1 | - | 2 - 3 | 257 | 94 | ||
FQ228978.1 | - | 2 - 3 | 489 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |