Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031820.1 | NP_114008.1 | 11 - 12 | 1213 | 77 | |
XM_008764388.1 | XP_008762610.1 | 10 - 11 | 1615 | 77 | ||
XM_008764390.1 | XP_008762612.1 | 10 - 11 | 1615 | 77 | ||
XM_008764391.1 | XP_008762613.1 | 11 - 12 | 1316 | 77 | ||
GenBank mRNA | AF143545.2 | - | 11 - 12 | 1213 | 77 | |
AF143546.2 | - | 11 - 12 | 1213 | 77 | ||
BC062070.1 | - | 7 - 8 | 1475 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |